

Acetylacetone, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:123-54-6
:
Formula:CH₃COCH₂COCH₃
MDL Number:MFCD00008787
:100.12 g/mol
Merck Index:13,00083
Boiling Pt:140.4 °C (1013 hPa)
Melting Pt:–23 °C
Density:0.96945 g/cm³ (20 °C)
EINECS:204-634-0
UN:2310
ADR:3,III
:M-Clarity™ quality level = MQ100
Flash Pt:30 °C
:GHS02
:GHS07
Storage Temperature:Ambient