

3-Pentanone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:96-22-0
:
Formula:C₅H₁₀O
MDL Number:MFCD00009320
:86.13 g/mol
Merck Index:13,03148
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–40 °C
Density:0.8148 g/cm³ (20 °C)
EINECS:202-490-3
UN:1156
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:13 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:50 g/l (20 °C)
- Melting Point:-40 °C
- Molar Mass:86.13 g/mol
- Boiling Point:101 - 103 °C (1013 mbar)
- Vapor Pressure:37.6 hPa (20 °C)
- Flash Point:7 °C
- Refractive Index:1.3923 (20 °C, 589 nm)
- Explosion Limit:1.6 - 7.7 %(V)
- Density:0.81 g/cm3 (20 °C)
- pH:6.2 (50 g/l, H2O, 20 °C)
- Ignition Point:425 °C
- Synonyms:3-Pentanone, Ethyl ketone
- Hill Formula:C5H10O