Order Entry
Sweden
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, 2-Pentanone
  46211-5ML
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, 2-Pentanone
  46211-5ML
 :  Merck
 :  

 

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    107-87-9
  • Description:
    2-Pentanone
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H302 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00009400
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Methyl propyl ketone
  • Melting point:
    –78 °C (lit.)
  • Formula:
    CH₃COCH₂CH₂CH₃
  • Molecular weight:
    86,13
  • Density:
    0,809 g/ml at 25 °C (lit.)
  • Flash point:
    7 °C - Closed cup
  • Boiling point:
    101 - 105 °C (lit.)

 

 

  • GHS signal: GHS02, GHS07
 : P210 - P233 - P240 - P241 - P301 + P312 - P305 + P351 + P338