Trifluoroacetic acid 0.1% (v/v) in aqueous solution
Catalog # ROTHCP05.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
About this item
:
:114.02 g/mol
Storage Temperature:Ambient
EINECS:200-929-3
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169