Order Entry
Sweden
ContactUsLinkComponent
Hydrochloric acid 1 mol/l (1 N) Reag. Ph. Eur. 3001800
Hydrochloric acid 1 mol/l (1 N) Reag. Ph. Eur. 3001800
Catalog # REGE3001800
Supplier:  REAGECON
CAS Number:  
Hydrochloric acid 1 mol/l (1 N) Reag. Ph. Eur. 3001800
Catalog # REGE3001800
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
ADR:8,II
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324