Saltsyra 10%
Katalog # REGEH10WV1
Supplier: REAGECON
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:1 L
- Seq:5037
Specifications
About this item
:
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS07
Storage Temperature:Omgivande
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324