Methanol
Catalog # 4.80467.9030
Supplier: Merck
CAS Number:
Specifications
- Pk:30 L
- Seq:5352
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
About this item
:
:32.04 g/mol
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
:GHS02
:GHS06
:GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595