Magnesium hydroxide carbonate heavy (Parteck® Mg DC), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, E504, SAFC®
:12125-28-9
:MFCD00149783
:MgCO₃ (linear)
:84.31 g/mol
:2.16 g/cm³ (20 °C)
:Ambient
:235-192-7
- Assay (complexometric calculated as Magnesiumoxide):40.0 - 43.5 %
- Assay USP (alkalimetric calculated as Magnesiumoxide):40.0 - 43.5 %
- Assay JP (complexometric calculated as Mangesiumoxide):40.0 - 44.0 %
- Identity:passes test
- Appearance of solution:passes test
- Substances soluble in water:≤ 1.0 %
- soluble salts:≤ 1.0 %
- Substances insoluble in acetic acid:≤ 0.05 %
- Substances insoluble in acid:≤ 0.05 %
- Chloride (Cl):≤ 0.02 %
- Nitrate (NO₃):≤ 0.015 %
- Nitrite (NO₂):≤ 0.015 %
- Sulfate (SO₄):≤ 0.3 %
- Heavy metals (as Pb):≤ 0.001 %
- As (Arsenic):≤ 0.0002 %
- Ca (Calcium):≤ 0.4 %
- CaO (Calcium oxide):≤ 0.6 %
- Fe (Iron):≤ 0.005 %
- Hg (Mercury):≤ 0.0001 %
- Na (Sodium):≤ 0.4 %
- Pb (Lead):≤ 0.0002 %
- Residual solvents (ICH Q3C):excluded by manufacturing process
- Bulk density:≥ 0.25 g/ml
- Particle Size (d50):21 - 51 µm
- Particle Size (d90):41 - 91 µm
- Precipitation test:≤ 12 ml
- Total aerobic microbial count:≤ 2000 CFU/g
- Total yeast and mould count:≤ 200 CFU/g
- E.coli (absent in 1 g):passes test