Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acetic acid
Acetic acid 4% (v/v) in aqueous solution
Acetic acid 4% (v/v) in aqueous solution
Acetic acid 4% (v/v) in aqueous solution
Leverantör:
Thermo Fisher Scientific
Certifikat
Expand image
Acetic acid 4% (v/v) in aqueous solution
Leverantör:
Thermo Fisher Scientific
Certifikat
Om denna produkt
CAS-nummer:
64-19-7
:
Formel:
H₃CCOOH
MDL-nummer:
MFCD00036152
:
60.05 g/mol
Förvaringstemperatur:
Ambient
EINECS:
200-580-7
Relaterad information
Documentation
Certifikat
Sök efter LOT-nummer
SDS
msds_ALFA035572
Videor
Thermo Scientific Acroseal 25 Years
Documentation