Order Entry
Sweden
ContactUsLinkComponent
4-Nitrophenyl acetate, Sigma-Aldrich®
4-Nitrophenyl acetate, Sigma-Aldrich®
Catalog # 8.43075.0025
Supplier:  Merck
CAS Number:  
4-Nitrophenyl acetate, Sigma-Aldrich®
Catalog # 8.43075.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    0.53 g/l (25 °C)
  • Melting Point:
    77 - 79 °C
  • Molar Mass:
    181.15 g/mol
  • Flash Point:
    110 °C
  • Hill Formula:
    C8H7NO4

Specification Test Results

About this item

 :
 :181.15 g/mol
Melting Pt:76…79 °C
 :M-Clarity™ quality level = MQ200
EINECS:212-593-5
CAS Number:830-03-5
Formula:C₈H₇NO₄
MDL Number:MFCD00007326