2-Ethoxybenzamide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:938-73-8
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00007977
:165.19 g/mol
Boiling Pt:333 °C (1013 hPa)
Melting Pt:129…134 °C
EINECS:213-346-4
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Melting point:129 to 134 °C
- HPLC:≥96.0 %
- Ferro compounds:≤0.05 % as ferrocyanide ion ¢Fe(CN)6!4-
- Insoluble matter:≤0.005 %