1-Hexanesulphonic acid sodium salt ≥98.0% (by titrimetric analysis) for ion pair chromatography
Supplier: TCI
Certificates
About this item
CAS Number:2832-45-3
:MFCD00007542
Formula:C₆H₁₃O₃SNa
Melting Pt:300 °C
:188.22 g/mol
Storage Temperature:Ambient
EINECS:220-601-3
Specification Test Results
- Appearance:White powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
- Absorbance (210nm):max. 1.000
- Absorbance (230nm):max. 0.300
- Absorbance (254nm):max. 0.130
Specification Test Results
Frequently Bought Together