Order Entry
Sweden
ContactUsLinkComponent
 
Organic reference standard, 2-Bromophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA10736200ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2-Bromophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA10736200ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    95-56-7
  • Description:
    2-Bromophenol 100 µg/ml in methanol
  • Formula:
    C₆H₅BrO
  • Description:
    2-Bromophenol

Specifications

About this item