Order Entry
Sweden
ContactUsLinkComponent
Organic reference standard, 2,3,4,5-Tetrachloroaniline 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, 2,3,4,5-Tetrachloroaniline 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17330400CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2,3,4,5-Tetrachloroaniline 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17330400CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    634-83-3
  • Description:
    2,3,4,5-Tetrachloroaniline 10 µg/ml in cyclohexane
  • Formula:
    C₆H₃Cl₄N
  • Description:
    2,3,4,5-Tetrachloroaniline

Specifications

About this item