Order Entry
Sweden
ContactUsLinkComponent
Acetonitrile, extra dry 99.9+%, AcroSeal®
Acetonitrile, extra dry 99.9+%, AcroSeal®
   Hållbara alternativ tillgängliga
Certifikat
undefined
Acetonitrile, extra dry 99.9+%, AcroSeal®
   Hållbara alternativ tillgängliga
Specifikationer
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Specifikationer
Specifikationstestresultat
  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.001 % (Coulometric)
  • GC:
    ≥99.90 %
  • Residue after evaporation:
    ≤0.0005 %
  • Acidity (CH3COOH):
    ≤0.002 %
  • Acetamide:
    ≤10 ppm
Specifikationstestresultat