Order Entry
Sweden
ContactUsLinkComponent
Double burette clamps
Double burette clamps
Catalog # 241-0350
Supplier:  Brand
undefined
Double burette clamps
Catalog # 241-0350
Supplier:  Brand
Specifications
  • For:
    Two burettes up to 50 ml
  • Pk:
    1
Specifications
About this item
Aluminium, PVC coated.
  • For two burettes up to 50 ml
  • PVC-coated clamps provide a secure grip
  • Comfortable height adjustment
  • Graduations remain visible
  • Very sturdy design, easy to operate
  • Integral support connector