Order Entry
Sweden
Orders LinkContactUsLinkComponent
Formaldehyde 37% in aqueous solution

Formaldehyde 37% in aqueous solution

200 L
Catalog # SWED52536-218
Toxic
Corrosive
Health
Supplier:   SWED HANDLING
Formaldehyde 37% in aqueous solution
Catalog # SWED52536-218
Supplier:  SWED HANDLING
CAS Number:  50-00-0
Formaldehyde 37% in aqueous solution

200 L

CAS Number:  50-00-0
Price
Inquire For Price
Log in for availability
1 * 200 L
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Merck Index:13,04259
  • Boiling Pt:–19.3 °C (1013 hPa)
  • Melting Pt:–92 °C
  • Density:0.8153 g/cm³ (–20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2209
  • ADR:8,III
Specifications

Specifications

  • Pk:
    200 L

Specifications