Order Entry
Sweden
ContactUsLinkComponent
1-Bromopropane for synthesis, Sigma-Aldrich®
Flammable
Health
Irritant
1-Bromopropane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Flammable
Health
Irritant
1-Bromopropane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    2.5 g/l (20 °C)
  • Melting Point:
    -108 °C
  • Molar Mass:
    122.99 g/mol
  • Boiling Point:
    71 °C (1013 hPa)
  • Vapor Pressure:
    146 hPa (20 °C)
  • Flash Point:
    -10 °C
  • Refractive Index:
    1.4341 (20 °C, 589 nm)
  • Explosion Limit:
    3.4 - 9.1 %(V)
  • Density:
    1.35 g/cm3 (20 °C)
  • Ignition Point:
    490 °C

Specification Test Results

Related Information