Order Entry
Sweden
ContactUsLinkComponent
 
 
 
 
1-Brompropan 99%
 
undefined
 
 
 
1-Brompropan 99%

 

  • Appearance:
    Clear colorless to light yellow Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4320 to 1.4360 (20°C, 589 nm)
  • GC:
    ≥98.5 %