Tin(II) chloride dihydrate ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 10025-69-1 Molecular Formula: Cl2Sn.2H2O Formula Weight: 225.65 (189.62anhy) Melting Point: 42-46° Merck Reference: 14,8783 MDL No.: MFCD00149863
Synonyms: Stannous chloride dihydrate
- CAS Number:10025-69-1
- Formula:SnCl₂·2H₂O
- MDL Number:MFCD00149863
- Molecular Weight:225.65 g/mol
- Merck Index:14,8783
- Boiling Pt:623 °C (1013 hPa)
- Melting Pt:42…46 °C
- Density:2.71
- Storage Temperature:Ambient
- EINECS:600-045-1
- UN:3260
- ADR:8,III
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 10025-69-1 Molecular Formula: Cl2Sn.2H2O Formula Weight: 225.65 (189.62anhy) Melting Point: 42-46° Merck Reference: 14,8783 MDL No.: MFCD00149863
Synonyms: Stannous chloride dihydrate
- CAS Number:10025-69-1
- Formula:SnCl₂·2H₂O
- MDL Number:MFCD00149863
- Molecular Weight:225.65 g/mol
- Merck Index:14,8783
- Boiling Pt:623 °C (1013 hPa)
- Melting Pt:42…46 °C
- Density:2.71
- Storage Temperature:Ambient
- EINECS:600-045-1
- UN:3260
- ADR:8,III
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



