

2-Iodothiophene ≥98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3437-95-4
:
Formula:C₄H₃IS
MDL Number:MFCD00005424
:210.04 g/mol
Boiling Pt:182 °C
Melting Pt:–40 °C
Density:2.064
Flash Pt:71 °C (159 °F)
:Stabilised with copper
Storage Temperature:Ambient
EINECS:222-342-1
:GHS06
:GHS07