

Calcium nitrate hydrate ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35054-52-5
:
Formula:Ca(NO₃)₂·H₂O
MDL Number:MFCD00149604
:182.1 g/mol
Merck Index:14,1682
Boiling Pt:132 °C (dec.)
Melting Pt:42 °C
Density:1.82
Storage Temperature:Ambient
EINECS:233-332-1
UN:1454
ADR:5.1,III
:GHS03
:GHS07
Frequently Bought Together