Sodium thiosulphate pentahydrate 99.5+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10102-17-7
:
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186
:248.19 g/mol
Merck Index:12,08844
Melting Pt:48.5 °C
Density:1.72 g/cm³
Storage Temperature:Ambient
EINECS:231-867-5
Specification Test Results
- Appearance:White Crystals
- Titration Iodimetric:99.5 to 101.0 %
- pH:6.0 to 8.4 (5 % solution at 25.0°C)
- Sulfide (S):Passes test (limit about 1 ppm)
- SO4 and SO3 (as SO4):≤0.1 %
- Insoluble matter:≤0.005 %
- Nitrogen compounds (as N):≤0.002 %