
Sodium dihydrogen phosphate monohydrate 98.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10049-21-5
:
Formula:NaH₂PO₄·H₂O
MDL Number:MFCD00149208
:137.99 g/mol
Merck Index:14,8660
Melting Pt:100 °C (-H₂O)
Density:2.04 g/cm³ (20 °C)
EINECS:231-449-2
:GHS07
Storage Temperature:Ambient