

Lithium hydroxide monohydrate (≥56.5% LiOH)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1310-66-3
:
Formula:LiOH·1H₂O
MDL Number:MFCD00149772
:41.96 g/mol
Merck Index:14,5534
Boiling Pt:920 °C (1013 hPa)
Melting Pt:462 °C
Density:1.51
Storage Temperature:Ambient
EINECS:215-183-4
UN:2680
ADR:8,II
:GHS07
:GHS05