di-Sodium tetraborate decahydrate 99.0-103.0% Ph. Eur.
Leverantör: VWR Chemicals
Certifikat
Om denna produkt
CAS-nummer:1303-96-4
:
Formel:Na₂B₄O₇·10H₂O
MDL-nummer:MFCD00149193
:381.37 g/mol
Merck Index:13,08662
Kokande Pt:320 °C (1013 hPa)
Smält Pt:75 °C
Densitet:1.73 g/cm³ (20 °C)
Förvaringstemperatur:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
:GHS08
Specifikationstestresultat
- Assay:99.0 - 103.0 %
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification B:Passes test
- Identification (Na):Passes test
- Appearance of solution:Passes test
- pH (4 %):9.0 - 9.6
- SO4 (Sulphate):≤ 50 ppm
- NH4 (Ammonium):≤ 10 ppm
- Ca (Calcium):≤ 100 ppm
- Heavy metals (as Pb):≤ 25 ppm
- Residual solvents:Passes test
- As (Arsenic):≤ 5 ppm
- Cd (Cadmium):≤ 1 ppm
- Co (Cobalt):≤ 1 ppm
- Hg (Mercury):≤ 2 ppm
- Ni (Nickel):≤ 1 ppm
- Pb (Lead):≤ 2 ppm
- V (Vanadium):≤ 1 ppm
- Conforms to Ph.Eur.:Passes test
Specifikationstestresultat
Relaterad information
Certifikat
Sök efter LOT-nummer2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 30L Plastic bucket UN Z (ZPACK13247037)
Packaging Information
83555_DI SODIUM TETRABORATE 10H2O PH.EUR_BSE_REV1
BSE/TSE Certificates
spec_VWRC83555.290 spec_VWRC83555.290 spec_VWRC83555.460 spec_VWRC83555.460
Product specification
VWR Chemicals for Biopharma and Pharma Production