Order Entry
Sweden
ContactUsLinkComponent
di-Sodium tetraborate decahydrate ≥97%, powder, TECHNICAL
Health
di-Sodium tetraborate decahydrate ≥97%, powder, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Health
di-Sodium tetraborate decahydrate ≥97%, powder, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 97 %

Specification Test Results

Frequently Bought Together