Sulphuric acid 0.5 M H2SO4 (1.0N) in aqueous solution ≥99.9% Reag. Ph. Eur. volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Boiling Pt:100 °C (1.013 hPa)
Melting Pt:<0 °C
Density:1.030 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-755-4
UN:3264
ADR:8,III
:GHS05
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective