(±)-1,2-Propanediol 99%, extra pure
:57-55-6
:
:CH₃CH(OH)CH₂OH
:MFCD00064272
:76.1 g/mol
:13,07947
:187 °C (760 mmHg)
:–60 °C
:1.03 g/cm³
:99 °C
:Ambient
:200-338-0
- Appearance:Clear viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4310 to 1.4330 (20°C, 589 nm)
- Water:≤0.2 % (Coulometric)
- GC:≥98.5 %
- Color scale:≤10 APHA