Order Entry
Sweden
ContactUsLinkComponent
Orthophosphoric acid ≥85%
Orthophosphoric acid ≥85%
Catalog # SWEDFO861302
Supplier:  SWED HANDLING
CAS Number:  
Orthophosphoric acid ≥85%
Catalog # SWEDFO861302
Supplier:  SWED HANDLING
CAS Number:  

Specifications

  • Pk:
    43 kg
  • Seq:
    4379

Specifications

About this item

 :
 :98 g/mol
Melting Pt:28 °C
Density:1.71…1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340