Order Entry
Sweden
Orders LinkContactUsLinkComponent
Sodium hydroxide 35% in aqueous solution, TECHNICAL

Sodium hydroxide 35% in aqueous solution, TECHNICAL

Supplier:   VWR Chemicals
Sodium hydroxide 35% in aqueous solution, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Sodium hydroxide 35% in aqueous solution, TECHNICAL
Sodium hydroxide 35% in aqueous solution, TECHNICAL

4 Options Available Below

Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications for Product Family

Specification Test Results

  • Assay:
    Min. 34 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Fe (Iron):
    Max. 50 ppm
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.