Order Entry
Sweden
ContactUsLinkComponent
Sodium hydroxide 32% in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Corrosive
Sodium hydroxide 32% in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Corrosive
Sodium hydroxide 32% in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (acidimetric):
    min. 32 %
  • Chloride (Cl):
    max. 0.002 %
  • Sodium Carbonate (acidimetric):
    max. 1 %
  • Silica (SiO2):
    max. 0.003 %
  • Sulfate (SO4):
    max. 0.001 %
  • Trace Impurities (in ppm) Aluminium (Al):
    max. 5
  • Trace Impurities (in ppm) Iron (Fe):
    max. 5
  • Trace Impurities (in ppm) Heavy Metals (as Pb):
    max. 5
  • Trace Impurities (in ppm) Phosphate (PO4):
    max. 5
  • Trace Impurities (in ppm) Total Nitrogen (N):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together