Hydrochloric acid 5 mol/l (5 N) volumetric solution
Catalog # REGEH25005
Supplier: REAGECON
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:5 L
- Seq:5190
Specifications
About this item
:
Density:1.06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,II
:36.46 g/mol
UN:1789
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324