Hydrochloric acid 37%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,II
:36.46 g/mol
REACH:01-2119484862-27
:GHS05
:GHS07
Specification Test Results
- Assay:35.0 - 38.0 %
- Appearance:Clear colourless liquid
- Appearance of solution:Passes test Ph.Eur.
- Identification A:Passes test Ph.Eur.
- Identification B:Passes test Ph.Eur.
- Colouration:Max. 10 APHA
- Density (20/4):1.174 - 1.193
- Evaporation residue:Max. 100 ppm
- Free chlorine:Max. 1 ppm
- Heavy metals (as Pb):Max. 2 ppm
- Ignition residue (SO4):Max. 5 ppm
- Reducing substances (as O):Max. 4 ppm
- Br (Bromide):Max. 5 ppm
- NH4 (Ammonium):Max. 1 ppm
- PO4 (Phosphate):Max. 0.5 ppm
- SO3 (Sulphite):Max. 1 ppm
- SO4 (Sulphate):Max. 1 ppm
- Al (Aluminium):Max. 0.2 ppm
- As (Arsenic):Max. 0.01 ppm
- Ba (Barium):Max. 0.02 ppm
- Be (Beryllium):Max. 0.01 ppm
- Bi (Bismuth):Max. 0.02 ppm
- Ca (Calcium):Max. 1 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cr (Chromium):Max. 0.07 ppm
- Cu (Copper):Max. 0.1 ppm
- Fe (Iron):Max. 0.3 ppm
- Ge (Germanium):Max. 0.02 ppm
- Hg (Mercury):Max. 0.01 ppm
- K (Potassium):Max. 0.1 ppm
- Li (Lithium):Max. 0.01 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.01 ppm
- Mo (Molybdenum):Max. 0.02 ppm
- Na (Sodium):Max. 0.5 ppm
- Ni (Nickel):Max. 0.04 ppm
- Pb (Lead):Max. 0.02 ppm
- Sr (Strontium):Max. 0.01 ppm
- Ti (Titanium):Max. 0.02 ppm
- Tl (Thallium):Max. 0.02 ppm
- V (Vanadium):Max. 0.02 ppm
- Zn (Zinc):Max. 0.5 ppm
- Zr (Zirconium):Max. 0.02 ppm
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 1L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415111) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 2,5L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415250)
Förpackningsinformation
Mer
20252_HYDROCHLORIC ACID 37 %_BSE_REV0
BSE/TSE-certifikat
spec_VWRC20252.244 spec_VWRC20252.244 spec_VWRC20252.290 spec_VWRC20252.295 spec_VWRC20252.295
Produktspecifikation
Mer