Hydrochloric acid fuming 37%, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
EINECS:231-595-7
UN:1789
ADR:8,II
:36.46 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberMERP1.00314.2500_TDS_EN ProductPage_100314
Product specification
msds_100314