Order Entry
Sweden
ContactUsLinkComponent
Hydrochloric acid 3.571 mol/l (N/0.28), Titripur® volumetric solution, Supelco®
Hydrochloric acid 3.571 mol/l (N/0.28), Titripur® volumetric solution, Supelco®
Catalog # 1.13134.9010
Supplier:  Merck
CAS Number:  
undefined
Hydrochloric acid 3.571 mol/l (N/0.28), Titripur® volumetric solution, Supelco®
Catalog # 1.13134.9010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    PE/alu Titripac®
  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
Density:1.06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,
 :M-Clarity™ quality level = MQ100
 :36.46 g/mol
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324