Order Entry
Sweden
ContactUsLinkComponent
 
Methylamine 40% (w/w) in aqueous solution
Catalog # L00894.0F
CAS Number:  
Methylamine 40% (w/w) in aqueous solution
Catalog # L00894.0F
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL
  • Seq:
    2895

Specifications

About this item

CAS Number:74-89-5
 :
Formula:CH₅N
MDL Number:MFCD00008104
 :31.06 g/mol
Merck Index:14,6014
Boiling Pt:49 °C
Melting Pt:–38 °C
Density:0.899
Flash Pt:–14 °C (7 °F)
EINECS:200-820-0
UN:1235
ADR:3,II
 :GHS02
 :GHS06
 :GHS05
 :GHS07
Storage Temperature:Ambient