Order Entry
Sweden
ContactUsLinkComponent
Hydrogen peroxide 35% stabilised
Corrosive
Irritant
Hydrogen peroxide 35% stabilised
Catalog # ROTH9683.2
Supplier:  Roth Carl
CAS Number:  
Corrosive
Irritant
Hydrogen peroxide 35% stabilised
Catalog # ROTH9683.2
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.11 g/cm³ (20 °C)
ADR:5.1,II
 :34.01 g/mol
EINECS:231-765-0
UN:2014
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333