Order Entry
Sweden
ContactUsLinkComponent
Beakers, low or tall form, Borosilicate glass 3.3
  BRND91217
 :  Brand
 : 91217
undefined
Beakers, low or tall form, Borosilicate glass 3.3
  BRND91217
 :  Brand
 : 91217
Select an Option
42 mm
Select an Option
50 ml
Select an Option
60 mm
Select an Option
10
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
  • Beakers 5 ml and 10 ml without graduations and labeling area
Volume graduations and labelling area.
 : DIN 12331, ISO 3819