Order Entry
Sweden
ContactUsLinkComponent
 
p-Xylene-[D₁₀]
Catalog # EHERC12370100
Supplier:  EHRENSTORFER
CAS Number:  
p-Xylene-[D₁₀]
Catalog # EHERC12370100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :116.09 g/mol
Boiling Pt:135 °C (1013 hPa)
Melting Pt:13,3 °C
Density:0,948 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:255-193-6
UN:1307
ADR:3,III
 :GHS02
 :GHS07
CAS Number:41051-88-1
Formula:C₆D₄(CD₃)₂
MDL Number:MFCD00037699