Methyl violet 2B
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 8004-87-3 Molecular Formula: C24H27N3.HCl Formula Weight: 393.96 Odour: Odourless MDL No.: MFCD00012568
Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
- CAS Number:8004-87-3
- Formula:C₂₄H₂₈ClN₃
- MDL Number:MFCD00012568
- Molecular Weight:393.96 g/mol
- Melting Pt:137 °C
- Storage Temperature:Ambient
- UN:3077
- ADR:9,III
2 Options Available Below
Product Details & Documents
CAS No.: 8004-87-3 Molecular Formula: C24H27N3.HCl Formula Weight: 393.96 Odour: Odourless MDL No.: MFCD00012568
Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
- CAS Number:8004-87-3
- Formula:C₂₄H₂₈ClN₃
- MDL Number:MFCD00012568
- Molecular Weight:393.96 g/mol
- Melting Pt:137 °C
- Storage Temperature:Ambient
- UN:3077
- ADR:9,III
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



