
Giemsa's stain, pure, high purity biological stain
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51811-82-6
:
MDL Number:MFCD00081642
Melting Pt:300 °C
EINECS:257-438-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark green to black Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤10 % (110°C)
- UV Lambda max:640 to 650 nm (in MeOH)
- Specific absorbance A (1%/1cm):≥950 (in MeOH) On dried substance