Orange G, pure, certified
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10
- CAS Number:1936-15-8
- Formula:C₁₆H₁₀N₂Na₂O₇S₂
- MDL Number:MFCD00012457
- Molecular Weight:452.38 g/mol
- Melting Pt:141 °C
- Density:0,45 to 0.47 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:217-705-6
2 Options Available Below
Product Details & Documents
Synonyms: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10
- CAS Number:1936-15-8
- Formula:C₁₆H₁₀N₂Na₂O₇S₂
- MDL Number:MFCD00012457
- Molecular Weight:452.38 g/mol
- Melting Pt:141 °C
- Density:0,45 to 0.47 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:217-705-6
Documents
ACRO41655
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Orange Powder
- Infrared spectrum:Conforms
- Dye content:≥80 %
- UV Lambda max:476 to 481 nm
- Absorption Ratio P-15/P+15:0.88 to 0.95
- BSC Certification:Passes
- Microbial contamination:≤10 /g (Escherichia coli)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



