Resazurin sodium salt
Catalog # TINT501800
Supplier: Lovibond Tintometer
CAS Number:
Specifications
- Pk:100 Tab.
- Seq:0940
Specifications
About this item
:
:251.17 g/mol
EINECS:263-718-5
CAS Number:62758-13-8
Formula:C₁₂H₆NNaO₄
MDL Number:MFCD00005036