Order Entry
Sweden
Orders LinkContactUsLinkComponent
Schiff reagent for analysis of aldehydes, Sigma-Aldrich®

Schiff reagent for analysis of aldehydes, Sigma-Aldrich®

Glass bottle, 500 mL
Catalog # 3952016-500ML
Corrosive
Health
Schiff reagent for analysis of aldehydes, Sigma-Aldrich®
Catalog # 3952016-500ML
CAS Number:  
Schiff reagent for analysis of aldehydes, Sigma-Aldrich®
Schiff reagent for analysis of aldehydes, Sigma-Aldrich®

Glass bottle, 500 mL

CAS Number:  
Price
961,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

Intended for use in Sigma Periodic Acid Staining Procedure, 395. Kit is intended for use with blood, bone marrow, tissue touch preparations or routine tissue sections.

Components: Pararosaniline, 1% and sodium metabisulfate, 4%, in hydrochloric acid, 0.25 mol/L

Use Sigma reagent S5133 for staining glycoproteins in polyacrylamide gels.

  • MDL Number:MFCD00081985
  • Density:1.1 g/cm³ (20 °C)
  • UN:1789
  • ADR:8,III
  •  :M-Clarity™ quality level = MQ500 (more info.)

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.