Analogue stopwatches, Amigo
: HANHART
Mechanical stopwatches with ABS casing.
- Winder on the back with lanyard
- Shock-protected pin lever movement
- Robust ABS case with cord for hanging round the neck
- 30 or 60 minutes with increments of ¹/₅ seconds
- Three coloured push buttons: start/stop and reset