Canister and barrel pumps, PumpMaster
: BURKLE
Manual positive displacement pump for filling, emptying, and transferring liquids between canisters and barrels. Positive pressure is produced using a manual piston and forces the liquid out of the container. Small quantities of liquid can also be drained off with the drain cock. The pump is fitted onto the barrel with a flexible, variable diameter plug. No special thread adapters are needed.
- Fits container openings with diameter from 49,5 to 60 mm
- Length of immersion tube 950 mm (immersion tube can be shortened depending on the depth of the container)
- For mild, aqueous liquids (with EPDM seals)
: PumpMaster for acids and chemical liquids is not suitable for hydrochloric acid.