Repetitive pipette, HandyStep® S
612-3858
: Brand
- Description:HandyStep® S
- Pk:1
Ergonomic and durable repetitive pipette that allows quick and easy serial pipetting and, in conjunction with Brand PD-Tips (10 different sizes from 0,1 µl to 50 ml), providing up to 49 repetitive dispensings from a single aspiration. Also compatible with Combitips®, Combitips® plus and other standard repetitive tips.
- Easy tip mounting - tips are simply inserted from below
- Eject key for contact-free, safe tip ejection
- Superior ergonomic handling
- DE-M marking, with serial number
- 59 different volumes with 10 PD-Tip sizes
The HandyStep® S works on the positive displacement principle, making it ideal for routine and research applications, as well as for difficult liquids (viscous, high density or high vapour pressure liquids).
* Stroke setting = % of nominal volume
: Supplied with DE-M marking, performance certificate, operating manual, shelf/rack mount, three PD-Tips (0,1, 1 and 10 ml).