Order Entry
Sweden
ContactUsLinkComponent
Laboratory blenders, Seward Stomacher® 80
Laboratory blenders, Seward Stomacher® 80
Supplier:  Seward (FermionX)
Certificates
undefined
Laboratory blenders, Seward Stomacher® 80
Supplier:  Seward (FermionX)
Specifications
  • Catalog No:
  • 432-3155
  • 432-3110
  • Model:
  • Stomacher® 80
  • Stomacher® 80
  • Paddle speed:
  • 230 / 265 / 300 ±5
  • 230 / 265 / 300 ±5 min⁻¹
  • Timer:
  • 30 / 60 / 120 / continuous
  • 30 / 60 / 120 / continuous
  • W×D×H:
  • 290×320×180 mm
  • 290×320×180 mm
  • Weight:
  • 15 kg
  • 15 kg