Certified colour reference standards
About this item
For routine calibration of colorimeters and verification of test data.
Lovibond® certified colour reference materials are supplied with full traceability to internationally recognised standards. Each standard comes in a 500 ml bottle with a 12-month guarantee of colour stability and full certification including MSDS. Individual values may vary but these are always specified on the certificate of calibration supplied with each standard.
14 Options Available Below
Product Details & Documents
For routine calibration of colorimeters and verification of test data.
Lovibond® certified colour reference materials are supplied with full traceability to internationally recognised standards. Each standard comes in a 500 ml bottle with a 12-month guarantee of colour stability and full certification including MSDS. Individual values may vary but these are always specified on the certificate of calibration supplied with each standard.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Pack typePkAvailabilityPrice
- 633-0004ASTM Colour 1, (certified to ISO 17025)
- 633-0005ASTM Colour 3, (certified to ISO 17025)
- 633-0012Gardner Colour 2, (certified to ISO 17025)
- 633-0013Gardner Colour 5, (certified to ISO 17025)
- TINT133991Pt-Co 0 mg Pt/l, certified under ISO 9001 quality systemPt-Co 0 mg Pt/l, certified under ISO 9001 quality systemGlass bottle500 mLLog in for availability1 * 500 mL1 150,00 kr
- 633-0008Pt-Co 5 mg Pt/l, certified under ISO 9001 quality systemPt-Co 5 mg Pt/l, certified under ISO 9001 quality system500 mLLog in for availability1 * 500 mL3 600,00 kr
- 633-0009Pt-Co 10 mg Pt/l, certified under ISO 9001 quality systemPt-Co 10 mg Pt/l, certified under ISO 9001 quality system500 mLLog in for availability1 * 500 mL3 600,00 kr
- 633-0056Pt-Co 15 mg Pt/l, certified under ISO 9001 quality systemPt-Co 15 mg Pt/l, certified under ISO 9001 quality systemPlastic bottle500 mLLog in for availability1 * 500 mL3 600,00 kr
- 633-0010Pt-Co 30 mg Pt/l, certified under ISO 9001 quality systemPt-Co 30 mg Pt/l, certified under ISO 9001 quality system500 mLLog in for availability1 * 500 mL3 600,00 kr
- 633-0011Pt-Co 50 mg Pt/l, certified under ISO 9001 quality systemPt-Co 50 mg Pt/l, certified under ISO 9001 quality system500 mLLog in for availability1 * 500 mL3 600,00 kr
- 633-0059Pt-Co 100 mg Pt/l, certified under ISO 9001 quality systemPt-Co 100 mg Pt/l, certified under ISO 9001 quality systemPlastic bottle500 mLLog in for availability1 * 500 mL3 600,00 kr
- 633-0006Saybolt Colour 0, (certified to ISO 17025)
- 633-0007Saybolt Colour +12, (certified to ISO 17025)
- 633-0258AOCS-Tintometer® Colour set for colorimeter AF710-3AOCS-Tintometer® Colour set for colorimeter AF710-31Log in for availability1 * 1 Items25 125,00 kr